Learn More
Fisherbrand™ Cheesecloth Wipes
Shop All Fisherbrand Products
Click to view available options
Packaging Quantity:
60 Pk.
Unit Size:
Pack of 60
Description
- Meets all cleanliness specifications of USP (United States Pharmacopoeia) and Federal Specification CCC-G-101c
- Certified to be 100% pure cotton of reagent-grade quality and purity
- Can be used as a filtering material due to their inertness to harsh solvent
Specifications
Specifications
| Product Type | Wipes |
| Material | Cheesecloth |
| Packaging Quantity | 60 Pk. |