Learn More
Lovibond™ Checkit™ Comparator Single Parameter Test Kit
Product Code. 11725468
Click to view available options
Range:
0 to 1mg/L
4 to 10pH
5.2 to 6.8mg/L
6 to 7.6mg/L
Unit Size:
Each
Analytes:
Ammonia
Copper
pH
This item is not returnable. View return policy
Product Code. 11725468
Brand: Lovibond™ 147130
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Aqualytic™ Checkit™ Comparator Single Parameter Test Kit includes all materials and operating instructions needed for mobile and static water analysis of a single parameter and comes in a sturdy and handy plastic case.
  • Compact and lightweight
  • Provides the basis for a comprehensive and easy-to-use analysis system
  • Comparator Discs contain a continuous color scale for exact color matching between standard and sample
Specifications
Includes Comparator apparatus, color comparison discs, cells, stirring rod, reagents, instructions
No. of Tests 30
Quantity 1 Pack
Range 4 to 10pH
Type Checkit™ Comparator Single Parameter Test Kit