Learn More
Upchurch Scientific Check valve assembly inline 1/16 inch O.D. tubingPEEK(TM)1.5psi (0.1bar) crack pressure96µL
Brand:  Upchurch Scientific 02013-33
Check valve assembly inline 1/16 inch O.D. tubingPEEK(TM)1.5psi (0.1bar) crack pressure96µL
Product Code. 13072730
  • ONLINE EXCLUSIVE PRICE
    244.00€
    210.18€ / Each
    Save 33.82€
    13% Off
Only null left
In Stock