Learn More
Upchurch Scientific Check valve assembly inline 1/16 inch O.D. tubingPEEK(TM)1.5psi (0.1bar) crack pressure96µL
Brand: Upchurch Scientific 02013-33
Check valve assembly inline 1/16 inch O.D. tubingPEEK(TM)1.5psi (0.1bar) crack pressure96µL