Learn More
Bochem™ Chain Clamp
Click to view available options
Diameter (Metric):
50 to 160 mm
Length (Metric) Arm:
250 mm
Unit Size:
Each
Description
- Zincked steel
- Chain is silicone coated
Specifications
Specifications
| Quantity | 1 Ea. |
| Diameter (Metric) | 50 to 160 mm |
| Length (Metric) Arm | 250 mm |