missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CGS 15943

Product Code. 18084132
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18084132 50 mg 50mg
18003872 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18084132 Supplier R&D Systems™ Supplier No. 1699/50
  • ONLINE EXCLUSIVE PRICE
    311.00€
    294.00€ / 50mg
    Save 17.00€
    5% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent adenosine receptor antagonist

CGS 15943 is a potent adenosine receptor antagonist (Ki values are 3.5, 4.2, 16 and 51 nM for human A1, A2A, A2B and A3 receptors respectively). Inhibits the catalytic subunit of the class IB PI3K isoform p110γ (IC50 = 1.1 μM). Blocks HCC and HDAC cell proliferation in vitro via inhibition of the PI3K/Akt pathway; reduces proliferation of ER+ breast cancer cells. Also acts as a BMP-4 mimetic to stimulate osteogenic differentiation. Orally active in vivo.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104615-18-1
Molecular Formula C13H8ClN5O

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 285.69
Purity >99%
Molecular Weight (g/mol) 285.69
Chemical Name or Material CGS 15943
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.