missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CFL1 (Human) Recombinant Protein
Human CFL1 full-length ORF ( AAH11005, 1 a.a. - 166 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001072-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MASGVAVSDGVIKVFNDMKVRKSSTPEEVKKRKKAVLFCLSEDKKNIILEEGKEILVGDVGQTVDDPYATFAKMLPDKDCRYALYDATYETKESKKEDLVFIFWAPESAPLKSKMIYASSKDAIKKKLTGIKHELQANCYEGVKDRCTLAEKLGGSAVISLEGKPL
Specifications
AAH11005 | |
cofilin 1 (non-muscle) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CFL1 | |
GST |
1072 | |
Wheat germ expression system | |
10 μg | |
CFL | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |