missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies CFDA-SE

Product Code. 17190352
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17190352 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17190352 Supplier STEMCELL Technologies Supplier No. 75003
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Cell proliferation and tracking dye

  • CFDA-SE (5(6)-carboxyfluorescein diacetate succinimidyl ester) is a stable, cell-permeable diacetate precursor to CFSE
  • Upon diffusion into the cell, intracellular esterases cleave the acetate group to generate CFSE, which interacts with cellular amines via its succinimidyl groups to generate a highly fluorescent green dye that is impermeant to the cell membrane
  • CFDA-SE is frequently used in cell proliferation assays, as it is partitioned approximately equally between the progeny so that cell division can be followed as a successive halving of the fluorescence intensity through multiple generational divisions
  • CFDA-SE is also used for motility assays and in vivo cell tracking experiments
  • CFDA-SE-labeled cells can be detected with any instrument/filter set compatible with fluorescein detection

Specifications

Quantity 10 mg
For Use With (Application) Flow Cytometry, Immunocytochemistry, Immunofluorescence
Cell Type Other
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.