Learn More
Thermo Scientific™ CETAC™ Autosampler Sample Probes
Product Code. 13211400 Shop All Thermo Scientific Products
Encompass
Change view
Click to view available options
Includes:
Sample Probe (0.5mm)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes Unit Size
13211400 Sample Probe (0.5mm) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 13211400 Supplier Thermo Scientific™ Supplier No. 942347003991
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
Choose a Thermo Scientific™ CETAC™ Sample Probe to suit your matrix and improve elemental analysis results with automated sample introduction with CETAC Autosamplers.
Customized autosampler accessories ensure more efficient, more reliable analysis with your Thermo Scientific™ CETAC™ autosampler and Trace Elemental Analyzers. Choose the appropriate CETAC Autosampler Sample Probe from this comprehensive range.
CETAC Standard Sample Probe
  • 0.5mm id
  • 0.8mm id
CETAC Carbon Fiber Sample Probe
Wider bore sample probe reduces the risk of blockage for aqueous samples that contain particulate matter
  • 0.5mm id
  • 0.8mm id
  • 1.0mm id for aqueous samples
CETAC Stainless Steel Filtered Probe for Oils
Designed especially for use with oil based samples, to ensure large particles are filtered out and avoid the risk of blockage.
Especificaciones
Type Autosampler Sample Probe
For Use With (Equipment) CETAC Autosampler
Includes Sample Probe (0.5mm)
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.