Learn More
Fisherbrand™ Certified Vial Kit 9mm short thread, Clear Glass, 1.5mL, PP Screw Cap, Center hole, Silicone/PTFE septum
Certified Kit, Includes 100 vials and 100 caps with pre-assembled septa.
Brand: Fisherbrand™ 11 40 2556
This item is not returnable.
View return policy
Description
- Certified Kit ND9
- 9mm
- Glass
- Clear
- 1.5mL
- Screw Cap
- Center hole (6 mm)
- Polypropylene
- Silicone/PTFE
- Reusable two compartment trays protect vials and closure while keeping matching supplies together