missing translation for 'onlineSavingsMsg'
Learn More

Cerium(IV) 2-methoxyethoxide, 18-20% w/w in 2-methoxyethanol

Product Code. p-7051108
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378438 5 g 5g
11398438 25 g 25g
11388438 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11378438 Supplier Thermo Scientific Alfa Aesar Supplier No. 040471.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 876107-33-4 | C12H28CeO8 | 440.464 g/mol

Hydrolysis of cerium 2-methoxyethoxide and subsequent supercritical drying or freeze-drying yielded a CeO 2 aerogel or cryogel with quite a large surface area above 300 m 2 /g.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 876107-33-4
Molecular Formula C12H28CeO8
Molecular Weight (g/mol) 440.464
MDL Number MFCD00236530
InChI Key JRRALIVAHBLZSS-UHFFFAOYSA-N
Synonym cerium iv methoxyethoxide w/w in methoxyethanol, tetrakis 2-methoxyethoxy cerium iv, cerium 4+ tetrakis 2-methoxyethan-1-olate, cerium iv methoxyethoxide in methoxyethanol
PubChem CID 21200429
IUPAC Name cerium(4+);2-methoxyethanolate
SMILES COCC[O-].COCC[O-].COCC[O-].COCC[O-].[Ce+4]

Specifications

Color Yellow
Density 1.02 g/mL
Linear Formula Ce[OCH2CH2OCH3]4
Quantity 5 g
UN Number UN1188
Sensitivity Moisture sensitive
Solubility Information Reacts with air.
Formula Weight 440.47
Concentration or Composition (by Analyte or Components) 18 to 20% w/w in 2-methoxyethanol
Physical Form Liquid
Chemical Name or Material Cerium(IV) 2-methoxyethoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.