missing translation for 'onlineSavingsMsg'
Learn More

Melford Cephalothin

Product Code. 30018187
Click to view available options
Quantity:
1 g
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 30018187

Brand: Melford C594501.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Cephalothin Sodium Salt is a first-generation cephalosporin antibiotic. It inhibits bacterial cell wall synthesis by binding to penicillin-binding proteins (PBPs).

This disruption interferes with the final stages of peptidoglycan synthesis in the bacterial cell wall, leading to structural instability and eventual cell lysis. Additionally, Cephalothin sodium salt is often used as a selective agent in culture media to isolate bacteria that are sensitive to this antibiotic. Its inclusion in culture medium allows for the specific growth of bacteria lacking resistance to cephalothin.

Specifications

Chemical Name or Material Cephalothin
CAS 58-71-9
Quantity 1 g
Molecular Formula C16H15N2NaO6S2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.