missing translation for 'onlineSavingsMsg'
Learn More

Cephalosporin 87/359(Cefoxitin Impurity H), TRC

Product Code. 30350944
Change view
Click to view available options
Quantity:
2.5 mg
Unit Size:
2.5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30350944 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30350944 Supplier Toronto Research Chemicals Supplier No. TRCC2431102.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cephalosporin 87/359(Cefoxitin Impurity H)
CAS 37051-05-1
Molecular Formula C15 H15 N3 O6 S2
Quantity 2.5 mg
Synonym (6R,7R)-3-[[(Aminocarbonyl)oxy]methyl]-8-oxo-7-[[2-(2-thienyl)acetyl]amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic Acid
InChI Formula InChI=1S/C15H15N3O6S2/c16-15(23)24-5-7-6-26-13-10(12(20)18(13)11(7)14(21)22)17-9(19)4-8-2-1-3-25-8/h1-3,10,13H,4-6H2,(H2,16,23)(H,17,19)(H,21,22)/t10-,13-/m1/s1
SMILES NC(=O)OCC1=C(N2[C@H](SC1)[C@H](NC(=O)Cc3cccs3)C2=O)C(=O)O
IUPAC Name (6R,7R)-3-(carbamoyloxymethyl)-8-oxo-7-[(2-thiophen-2-ylacetyl)amino]-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid
Molecular Weight (g/mol) 397.426
Formula Weight 397.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.