Learn More
Nickel Electro™ Fixed Angle Head Rotor for PCR Strips
Product Code. 12893272
Click to view available options
Capacity:
4 x 0.2 mL
Unit Size:
Each
Product Code. 12893272
Brand: Nickel Electro™ R0002
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Nickel Electro™ Fixed Angle Head Rotor for PCR Strips is comprised of 4 rows for 0.2mL PCR sprips with a max speed of 6000rpm.
  • Fixed
  • Angle head rotor
  • For PCR Strips
Specifications
Capacity 4 x 0.2 mL
Max. Speed 6,000 rpm