Learn More
OHAUS™ Centrifuge Adapter
Product Code. p-7085839 Shop All Ohaus Products
Click to view available options
Capacity:
15 x 1.5/2.0 mL
175/225 mL
250 mL
3 x 50 mL
4 x 30 mL
50 mL
6 x 15 mL
Description:
FA x2
FA x3
FB x2
MTB x2
RB x2
Bore Size (Metric):
Depth 11 mm
Depth 17 mm
Depth 25.5 mm
Depth 29 mm
Depth 29.5 mm
Depth 60 mm
Depth 60.5 mm
Depth 61 mm
Depth 61.5 mm
Unit Size:
Pack of 2
Pack of 3
Product Code. 16682784
Brand: OHAUS™ 30559419
Adapter accessory for Centrifuge Multi models
  • Adapter for various Centrifuge Multi models
  • Various capacities available
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Capacity 50 mL
For Use With (Equipment) Centrifuge Multi FC5916R 120 V and Centrifuge Multi FC5916 120 V
Bore Size (Metric) Depth 29.5 mm
Description FA x2