Learn More
Tork Centrefeed Wiping Paper M2
GREENER_CHOICE
Product Code. p-200113545 Shop All Tork Products
Click to view available options
Length (Metric):
275 m
Unit Size:
6 rolls
Product Code. 30243833
Brand: Tork 151131
Only null left
Add to basket
Edge
Add to basket
The 1-ply multipurpose Tork Wiping Paper is ideal for light wiping tasks and handwiping. This paper can be used in the Tork™ Centrefeed dispenser, which is a high capacity versatile solution for professional environments where both hand and surface wiping is required.
  • Ideal for wiping hands and lightly soiled surfaces
  • Unrestricted flow feature allows users to take as much as their tasks needs
  • Particularly good for cleaning glass - does not leave any lint or streaks on surfaces
  • Operable with one hand makes it easy to use
TRUSTED_SUSTAINABILITY
Specifications
Product Type Wiping Paper
Length (English) 300.74 yd.
Length (Metric) 275 m
Width (English) 8.46 in.
Width (Metric) 21.5 cm
Material Paper
Autoclavable Not Autoclavable
Certifications/Compliance EU Ecolabel, FSC Chain-Of-Custody, ISO 9001, ISO 14001, ISO 45001, ISO 50001, FSC Mix Virgin and Rec.fibre
Color White
Packaging Quantity 6/Pk.
Ply 1-ply
Certifications/Compliance (Europe) EU Ecolabel, FSC Chain-Of-Custody, ISO 9001, ISO 14001, ISO 45001, ISO 50001, FSC Mix Virgin and Rec.fibre