Learn More
R&D Systems™ CellXVivo Human Treg Cell Differentiation Kit
Product Code. 18431420
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18431420 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18431420 Supplier R&D Systems™ Supplier No. CDK006
  • ONLINE EXCLUSIVE PRICE
    531.00€
    501.90€ / 1 sets
    Save 29.10€
    5% Off
 Restricted Item
Estimated Shipment: 28-08-2026
Add to basket
Add to basket
For the differentiation of human regulatory T (Treg) cells from a preparation of naïve CD4+ T cells.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Includes Mouse Anti-Human CD3 Antibody
Human Treg Reagent 1
Human Treg Reagent 2
Human Treg Reagent 3
Human Treg Reagent 4
Reconstitution Buffer 1
Reconstitution Buffer 2
Wash Buffer (20X)
Quantity 1 Kit