Learn More
LABPLAS Cellulose Sani-Stick with Neutralizing Buffer
Product Code. 17946509
Change view
Click to view available options
Sterility:
Sterile
Unit Size:
Pack of 100
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Sterility Unit Size
17946509 Sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17946509 Supplier LABPLAS Supplier No. KSS67315NE
  • ONLINE EXCLUSIVE PRICE
    354.00€
    338.28€ / Pack of 100
    Save 15.72€
    4% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Cellulose sponge with stick for sampling surfaces, with buffer to neutralize disinfectant.
  • Cellulose Sani-Stick
  • Saturated with neutralizing buffer
  • Dimensions: 160 x 229 mm
  • Holds: 600 mL
  • Includes gloves
Specifications
Product Type Hygiene Sponge Sampling Kit
Media Type Neutralizers