Learn More
Macherey-Nagel™ Cellulose Glass Plates
Product Code. 10460702
Click to view available options
Dimensions (L x W):
20 x 20 cm
Unit Size:
Pack of 25
Product Code. 10460702
Brand: Macherey-Nagel™ 810053
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Specifications
Material Cellulose
Quantity 25 Pk.
Type Glass Plate
Thickness (Metric) 0.25 mm
Dimensions (L x W) 20 x 20 cm