Learn More
Macherey-Nagel™ Cellulose Extraction Thimbles
Click to view available options
Diameter (Metric) Inside:
15 mm
22 mm
31 mm
Height (Metric):
80 mm
100 mm
118 mm
180 mm
Unit Size:
Pack of 25
Description
- Manufactured from pure cellulose
- Used in the fields of air and waste gas analysis for collecting solid particles
- The choice of analytical chemists using Soxhlet or automated extractors
Specifications
Specifications
| Quantity | 25 Pk. |
| Diameter (Metric) Inside | 31 mm |
| Material | Cellulose |
| For Use With (Application) | Analysis of soil contaminants, Analysis of soil contaminants |
| Height (Metric) | 180 mm |
| Product Type | Extraction Thimbles |