Learn More
Advantec Mfs™ Cellulose Ester Membrane
Product Code. 11704799
Change view
Click to view available options
Pore Size:
0.45 μm
0.8 μm
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
11704799 0.8 μm Pack of 100
11789948 0.45 μm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11704799 Supplier Advantec Mfs™ Supplier No. A080H047A
MCE membranes are popular for many applications in the microbiological examination of water, wastewater, pharmaceuticals, food and beverages, and air monitoring. They offer high recovery rates and superior flow rates.
  • Mixed Cellulose Esters (MCE)
  • High recovery rates and superior flow rates
  • Contrasting gridlines facilitate counting colonies and are tested to assure freedom from grid line inhibition
  • EtO sterilised
  • Individually packaged
  • 47 mm Diameter
Spécification
Color Black
Filter Type Gridded Filter
Format Disc
Material Cellulose Ester
Pore Size 0.8 μm
Sterility Sterile
For Use With (Application) For capturing and culturing microorganisms using the MF Technique in the Water and Wastewater Industry
Includes No Pad
No. per Pack 100 Pk.
Sterilization Method EtO
Type Membrane Filter
Diameter (English) 1.9 in.
Diameter (Metric) 47 mm
Afficher plus Afficher moins
compliance-icons
Mention d'avertissement
  • Danger
Catégorie de danger
  • Flammable solid Category 1
  • Serious eye damage/eye irritation Category 2
Mention de danger
  • H228-Flammable solid.
Conseils de prudence
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P220-Keep/Store away from clothing/combustible materials.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
Informations supplémentaires
  • MIXTURE LIST-Contain: nitrocellulose
Nom du produit
Sélectionnez un problème
En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.