Learn More
Greiner Bio-One CELLSTAR 96-well, Non-Treated, Flat-Bottom Microplate
Click to view available options
Color:
Clear
Unit Size:
Pack of 60
Description
- Free of detectable DNase, RNase, human DNA
- Non-Pyrogenic
- Compatible with automated systems
- Alphanumeric well coding
- High clarity of microplates for optimal microscopic examinations
- Stackable
- Individu
Specifications
Specifications
| Format | Standard |
| Lid | With Lid |
| Array | 8 x 12 |
| Autoclavable | Not Autoclavable |
| Color | Clear |
| Material | Polystyrene |
| No. of Wells | 96 |
| Volume (Metric) Working | 25 to 340 μL |
| Sterility | Sterile |
| Surface Treatment | Non-treated |
| Show More |