Learn More
Corning™ cellgro™ Fetal Bovine Serum
GREENER_CHOICE
Product Code. 15323681
Change view
Click to view available options
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Unit Size
15323681 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15323681 Supplier Corning™ Supplier No. 35010CF
  • ONLINE EXCLUSIVE PRICE
    121.69€
    46.63€ / 50mL
    Save 75.06€
    61% Off
 Restricted Item
In Stock
The Corning range of high-quality serum products perfectly complement the Corning range of sterile-filtered, low-endotoxin, classical cell culture media. Within this portfolio, US-sourced Premium Fetal Bovine Serum, with the lowest endotoxin and haemoglobin specifications, is collected from cattle herds only within the United States. It is also available heat-inactivated. USDA Approved Source Fetal Bovine Serum, also low in endotoxin and haemoglobin, is collected from abattoirs located in countries free of BSE and FMD and approved by the United States Department of Agriculture (USDA) for bovine product import. The animals are inspected pre and post-mortem by licensed veterinarians and the material is tested for bovine diseases prior to import and further processing in FDA registered facilities under cGMPs. This is also available heat-inactivated. Specialty Serum products are also offered by Corning for a variety of product applications. All sera is separated from the collected blood by refrigerated centrifugation, true-pooled, filtered, and immediately frozen. Only raw serum that meets our strict specifications for endotoxin, residual haemoglobin and pH is approved for production. Fetal Bovine Serum (FBS) is a light brown-colored liquid separated from the blood of the bovine foetus. All material meets the approval of the U. S. Department of Agriculture and is obtained from sources free of BSE, Foot and Mouth Disease, and other reportable diseases pertaining to the species.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance USDA
Product Type Fetal Bovine Serum
For Use With (Application) Cell Culture
USDA Safety Testing Included USDA Inspected