Learn More
BD Biosciences™ Cell Viability Kit (RUO/GMP)
Product Code. 15746402
Change view
Click to view available options
Quantity:
100 Tests
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15746402 100 Tests 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15746402 Supplier BD Biosciences™ Supplier No. 349483
 Restricted Item
Estimated Shipment: 30-09-2026
Suitable for mammalian or microbial cell-based flow cyometry analyses.
  • Rapid counting of live/dead bacteria or other microbial cells.
  • Efficacy of bacterial disinfectant.
  • Viability of yeast during fermentation.
  • Viability and concentration of cells in culture.
  • Viability and concentration of cells before staining for flow cytometric analysis.
  • Mammalian or microbial cells.
  • Viability and count of cells in bioreactors.
  • Viability of sperm for research studiesiability in cell preparations containing debris.
  • Contents: Thiazole Orange solution, propidium iodide solution, liquid counting beads.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Thizole orange, propidium iodide
Includes The kit contains 1 vial of 500 μL 42 μmol/L TO in dimethyl sulfoxide (DMSO) and 1 vial of 500 μL 4.3 mmol/L PI in water.
No. of Tests 100 Tests
Quantity 100 Tests
Stability Stable for the period shon on the bottle label when stored as directed.