Learn More
BD Biosciences™ Cell Viability Kit with Liquid Counting Beads (RUO/GMP)
Description
- Rapid counting of live/dead bacteria or other microbial cells.
- Efficacy of bacterial disinfectant.
- Viability of yeast during fermentation.
- Viability and concentration of cells in culture.
- Viability and concentration of cells before staining for flow cytometric analysis.
- Mammalian or microbial cells.
- Viability and count of cells in bioreactors.
- Viability of sperm for research studiesiability in cell preparations containing debris.
- Contents: Thiazole Orange solution, propidium iodide solution, liquid counting beads.
Specifications
Specifications
| Content And Storage | Thizole orange, propidium iodide, liquid counting beads. |
| Includes | The kit contains 1 vial of 500 μL 42 μmol/L TO in dimethyl sulfoxide (DMSO) and 1 vial of 500 μL 4.3 mmol/L PI in water. Liquid Counting Beads are supplied as 1 vial of 10 mL of fluorescent microspheres in buffer with 0.1% sodium azide. |
| No. of Tests | 100 Tests |
| Quantity | 100 Tests |
| Stability | Stable for the period shon on the bottle label when stored as directed. |