Learn More
Abnova™ Cell Cycle Control Phospho-Specific Array
Description
- This product must be accompanied with Antibody Array Detection Kit
- The arrays are printed on standard-size coated glass microscope slides and can be scanned on all microarray scanners that are compatible with 76 x 25 x 1 mm slides
- Suitable Sample: Cultured Cells, Frozen Tissue, FFPE Tissue, Serum
Specifications
Specifications
| Content And Storage | 4°C |
| Detection Method | Fluorometric |
| Quantity | 1 Kit |
| For Use With (Application) | Profiling |
| Product Type | Array/Library |