Learn More
Corning™ Cell Culture Untreated Flasks with Vent Cap
BIOPHARMA
Product Code. 10783394 Shop All Corning Products
Change view
Click to view available options
Capacity (Metric):
10 mL
60 mL
210 mL
250 mL
Culture Area:
150 cm2
175 cm2
25 cm2
75 cm2
Volume (Metric) Working:
15 to 22.5 mL
30 to 45 mL
35 to 52.5 mL
5 to 7.5 mL
Unit Size:
Pack of 100
Pack of 200
Pack of 50
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Culture Area Volume (Metric) Working Unit Size
10783394 250 mL 175 cm2 35 to 52.5 mL Pack of 50
10399493 210 mL 150 cm2 30 to 45 mL Pack of 50
10125863 60 mL 75 cm2 15 to 22.5 mL Pack of 100
10258773 10 mL 25 cm2 5 to 7.5 mL Pack of 200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10783394 Supplier Corning™ Supplier No. 431466
  • ONLINE EXCLUSIVE PRICE
    355.00€
    331.29€ / Pack of 50
    Save 23.71€
    6% Off
In Stock
Printed with lot numbers for ease in traceability
  • Manufactured from optically clear virgin polystyrene
  • 100% integrity tested
  • Sterilized by gamma irradiation and certified nonpyrogenic
Cap Types
  • Phenolic-style screw caps for cultures requiring gas exchange
  • Plug seal caps for airtight cultures
  • Vent caps to minimize cell culture contamination with gas exchange
Canted-neck flasks feature:
  • Antitip skirt
  • Molded graduations
  • Frosted marking areas on both sides
  • Support bar across the top
  • Redesigned neck on 75cm2 flask features redesigned neck to aid media pouring, 1.4 in. (3.6cm) height for lower stacking height; vent cap has Costar-style concentric circle
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Orange
Closure Material HDPE
Capacity (Metric) 250 mL
Material Polystyrene
For Use With (Application) For optimal cell attachment
Includes Lot numbers for ease in traceability
Neck Style Angled
No. per Case 50/Cs.
No. per Pack 5/Pk.
Product Type Cell Culture Flask
Shape Rectangular
Surface Coating Not Treated
Writing Surface Yes
Graduated Yes
Sterility Sterile
Class Class I
Bottom Shape U-shape
Closure Type Vented
Culture Area 175 cm2
Surface Treatment Untreated
Volume (Metric) Working 35 to 52.5 mL
Closure Size 33 mm