Learn More
Cefixime, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 79350-37-1 |
| Molecular Formula | C16H15N5O7S2 |
| MDL Number | MFCD00865020 |
| Quantity | 100 mg |
| SMILES | O=C(O)C(N1[C@@]([H])([C@@H](C1=O)NC(/C(C2=CSC(N)=N2)=N\OCC(O)=O)=O)SC3)=C3C=C |
| Molecular Weight (g/mol) | 453.45 |
| Percent Purity | 97% |
| Chemical Name or Material | Cefixime |