Learn More
R&D Systems™ Cediranib
Product Code. 18786206
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18786206 10 mg 10mg
18708156 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18786206 Supplier R&D Systems™ Supplier No. 7454/10
 Restricted Item
Estimated Shipment: 30-10-2026
A Tocris product, part of R&D Systems. Potent inhibitor of VEGFR, PDGFR and FGFR
Cediranib is a highly potent and ATP-competitive inhibitor of VEGFR2 (IC50 = <1 nM). Cediranib also potently inhibits PDGFR1, VEGFR3, VEGFR1, c-Kit, PDGFR2 and FGFR1 (IC50 values are 2, 3, 5, 5, 26 and 36 nM, respectively), and inhibits CSF-1R, Src, and Abl (IC50 values are 110, 130 and 260 nM, respectively). Cediranib induces hypoxia, prevents VEGF-induced angiogenesis and suppresses homology-directed DNA repair (HDR) factors BRCA1/2 and RAD51. Cediranib decreases cell motility, proliferation and cell viability. Cediranib is active in vivo.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 4-[(4-Fluoro-2-methyl-1 H-indol-5-yl)oxy]-6-methoxy-7-[3-(1-pyrrolidinyl)propoxy]quinazoline
CAS 288383-20-0
Quantity 10 mg
Product Line Tocris
Target VEGF Receptor Inhibitors
Molecular Formula C25H27FN4O3
Purity 0.98