missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cedarwood Oil, Natural, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
:
500mL, Amber glass bottle
Specifications
Specifications
| CAS | 8000-27-9 |
| Color | Yellow |
| Boiling Point | 279°C |
| Packaging | Amber glass bottle |
| MDL Number | 132766 |
| Quantity | 500 mL |
| Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction