Description
Application Notes:This peptide is useful as a blocking peptide for NB600-284.
Specifications
Specifications
| Gene ID (Entrez) | 65266 |
| Species | Human |
| Product Line | Novus |
| Common Name | WNK4 |
| Concentration | 1 mg/mL |
| Content And Storage | Aliquot and store at -20 C or -80 C. Avoid freeze-thaw cycles. |
| Formulation | PBS |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB600-284. For Further Blocking Peptide Related Protocol, Click Here . |
| Gene Alias | EC 2.7.11, EC 2.7.11.1, PHA2B, PRKWNK4protein kinase, lysine deficient 4, Protein kinase lysine-deficient 4, Protein kinase with no lysine 4, serine/threonine-protein kinase WNK4, WNK lysine deficient protein kinase 4 |
| Gene Symbol | WNK4 |
| Show More |