missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CDKN2C Recombinant Protein

Brand:  Abnova™ H00001031-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16123071

  • 335.00€ / 10µg
Estimated Shipment: 29-05-2024
to see stock.
Add to basket

Description

Description

ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array

Specifications

Specifications

AAH16173
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
44.6
8
Glutathione Sepharose 4 Fast Flow
10 ug
MAEPWGNELASAAARGDLEQLTSLLQNNVNVNAQNGFGRTALQVMKLGNPEIARRLLLRGANPDLKDRTGFAVIHDAARAGFLDTLQTLLEFQADVNIEDNEGNLPLHLAAKEGHLRVVEFLVKHMASNVGHRNHKGDTACDLARLYGRNEVVSLMQANGAGGATNLQ
INK4C/p18/p18-INK4C
CDKN2C
Wheat Germ (in vitro)
GST
Antibody Production, Protein Array, ELISA, Western Blot
1031
CDKN2C (Human) Recombinant Protein (P01)
In vitro wheat germ expression system
12.5% SDS-PAGE Stained with Coomassie Blue.
Wheat Germ (in vitro)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDKN2C
Human
Recombinant
Solution
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers