Learn More
Abnova™ CDK4 293T Cell Transient Overexpression Lysate (Denatured)
Product Code. 16146104
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. 16146104
Brand: Abnova™ H00001019T02.100uL
Human overexpression lysate
Specifications
Cell Line 293T
For Use With (Application) Western Blot
Description CDK4 293T Cell Transient Overexpression Lysate(Denatured)
Gene Accession No. NM_000075.2
Molecular Weight (g/mol) Theoretical M.W. 33.7kDa
Plasmid pCMV-CDK4 full-length
Quality Control Testing Transient overexpression cell lysate was tested with anti-CDK4 antibody (Mfr. No. H00001019-D01P) by Western Blot.
Quantity 100 μL
Storage Buffer 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Host Species Human
Content And Storage Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Show More Show Less
compliance-icons
Product Identifier
  • 100UL CDK4 293T Cell Transient Overexpression Lysa
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Mercaptoethanol