missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CDK2 (Human) Recombinant Protein
Human CDK2 full-length ORF ( AAA35667, 1 a.a. - 298 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00001017-P02.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MENFQKVEKIGEGTYGVVYKARNKLTGEVVALKKIRLDTETEGVPSTAIREISLLKELNHPNIVKLLDVIHTENKLYLVFEFLHQDLKKFMDASALTGIPLPLIKSYLFQLLQGLAFCHSHRVLHRDLKPQNLLINTEGAIKLADFGLARAFGVPVRTYTHEVVTLWYRAPEILLGCKYYSTAVDIWSLGCIFAEMVTRRALFPGDSEIDQLFRIFRTLGTPDEVVWPGVTSMPDYKPSFPKWARQDFSKVVPPLDEDGRSLLSQMLHYDPNKRISAKAALAHPFFQDVTKPVPHLRL
Specifications
AAA35667 | |
cyclin-dependent kinase 2 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDK2 | |
GST |
1017 | |
Wheat germ expression system | |
10 μg | |
p33(CDK2) | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |