Learn More
CDK10 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA596192
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: MDKEKDGIPI SSLREITLLL RLRHPNIVEL KEVVVGNHLE SIFLVMGYCE QDLASLLENM PTPFSEAQVK CIVLQVLRGL QYLHRNFIIH RDLKVSNLLM TDKGCVKTAD FGLARAYGVP VKPMTPKVVT LWYRAPELLL GTTTQTTSID MWAVGCILAE LLAHRPLLPG TSEIHQIDLI VQLLGTPSEN IWPGFSKLPL VGQYSLRKQP YNNLKHKFPW LSEAGLRLLH FLFMATAGDC LESSYFKEKP LPCEPELMPT FPHHRNKRAA PATSEGQSKR CKP; Positive Samples: 293T, HT-29
Regulates, cooperatively with NONO and SFPQ, androgen receptor-mediated gene transcription activity in Sertoli cell line. Binds to poly(A), poly(G) and poly(U) RNA homopolymers.Specifications
CDK10 | |
Polyclonal | |
Unconjugated | |
CDK10 | |
CDK10 | |
Recombit fusion protein containing a sequence corresponding to amino acids 1-283 of human CDK10. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
2.9 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q15131, Q3UMM4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234854, 8558 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |