Learn More
Abnova™ CD96 Recombinant Protein
Human CD96 full-length ORF recombinant protein with GST-tag at N-terminal
Brand: Abnova™ H00010225-P01.10ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene belongs to the immunoglobulin superfamily. It is a type I membrane protein. The protein may play a role in the adhesive interactions of activated T and NK cells during the late phase of the immune response. It may also function in antigen presentation. Alternative splicing occurs at this locus and two transcript variants encoding distinct isoforms have been identified.
- Theoretical MW (kDa): 69.96
- Preparation method: In vitro wheat germ expression system
- Purification: Glutathione Sepharose 4 fast flow
- Storage buffer: 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer
Sequence: MEKKWKYCAVYYIIQIHFVKGVWEKTVNTEENVYATLGSDVNLTCQTQTVGFFVQMQWSKVTNKIDLIAVYHPQYGFYCAYGRPCESLVTFTETPENGSRWTLHLRNMSCSVSGRYECMLVLYPEGIQTKIYNLLIQTHVTADEWNSNHTIEIEINQTLEIPCFQNSSSKISSEFTYAWSVEDNGTQETLISQNHLISNSTLLKDRVKLGTDYRLHLSPVQIFDDGRKFSCHIRVGPNKILRSSTTVKVFAKPEIPVIVENNSTDVLVERRFTCLLKNVFPKANITWFIDGSFLHDEKEGIYITNEERKGKDGFLELKSVLTRVHSNKPAQSDNLTIWCMALSPVPGNKVWNISSGKITFLLGSEISSTDPPLSVTESTLDTQPSPASSVSPASKNVFTLSY
Best use within three months from the date of receipt of this protein.
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH20749 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
69.96 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
MEKKWKYCAVYYIIQIHFVKGVWEKTVNTEENVYATLGSDVNLTCQTQTVGFFVQMQWSKVTNKIDLIAVYHPQYGFYCAYGRPCESLVTFTETPENGSRWTLHLRNMSCSVSGRYECMLVLYPEGIQTKIYNLLIQTHVTADEWNSNHTIEIEINQTLEIPCFQNSSSKISSEFTYAWSVEDNGTQETLISQNHLISNSTLLKDRVKLGTDYRLHLSPVQIFDDGRKFSCHIRVGPNKILRSSTTVKVFAKPEIPVIVENNSTDVLVERRFTCLLKNVFPKANITWFIDGSFLHDEKEGIYITNEERKGKDGFLELKSVLTRVHSNKPAQSDNLTIWCMALSPVPGNKVWNISSGKITFLLGSEISSTDPPLSVTESTLDTQPSPASSVSPASKNVFTLSY | |
DKFZp667E2122/MGC22596/TACTILE | |
CD96 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
10225 | |
CD96 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD96 | |
Human | |
Recombinant | |
Solution |