Learn More
CD63 Rabbit anti-Human, CoraLite 594, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
129.00€ - 430.00€
Specifications
Antigen | CD63 |
---|---|
Applications | Flow Cytometry |
Classification | Polyclonal |
Conjugate | CoraLite 594 |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17781399
|
Proteintech
CL594-25682-20UL |
20 μL |
129.00€
20µL |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
17619956
|
Proteintech
CL594-25682-100UL |
100 μL |
430.00€
100µL |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
Description
CD63 Polyclonal antibody apecifically detetects CD63 antigen in Human Samples. It is validated for Flow Cytometry.Specifications
CD63 | |
Polyclonal | |
Liquid | |
Human | |
BC002349 | |
CD63 | |
IgG | |
Antigen affinity purification |
Flow Cytometry | |
CoraLite 594 | |
Rabbit | |
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
967 | |
CD63 fusion protein Ag19690 | |
Primary | |
Store at -20°C. Avoid exposure to light. |