missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD41 Rat anti-Mouse, PE, Clone: MWREG30, BD
Rat Monoclonal Antibody
151.00€ - 525.00€
Specifications
Antigen | CD41 |
---|---|
Clone | MWREG30 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15851399
|
BD Biosciences
561850 |
25 μg |
151.00€
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
15809178
|
BD Biosciences
558040 |
0.1 mg |
525.00€
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
Description
The MWReg30 antibody reacts with glycoprotein (gp) IIb, also called integrin αIIb chain (CD41), that associates with integrin β3 chain (gpIIIa or CD61) to form the gpIIb/IIIa (CD41/CD61) complex. CD41/CD61 is expressed on platelets, megakaryocytes, and early hematopoietic progenitors. The integrin complex binds to fibrinogen, fibronectin, vitronectin, von Willebrand factor, and thrombospondin. It is important for platelet adhesion and aggregation, and it may play a role in osteolytic tumor metastasis.
Specifications
CD41 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Mouse Platelets | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
MWREG30 | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
Integrin α IIb chain |
For Research Use Only.