missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD370 (Clec9A) Rat anti-Mouse, BB515, Clone: 10B4, BD Horizon™
Rat Monoclonal Antibody
Brand: BD Biosciences 566033
Additional Details : Weight : 0.01000kg
Description
CD370 (Clec9A) Monoclonal antibody specifically detects CD370 (Clec9A) in Mouse samples. It is validated for Flow cytometry.
CD370 (Clec9A) Monoclonal antibody specifically detects CD370 (Clec9A) in Mouse samples. It is validated for Flow cytometry.Specifications
CD370 (Clec9A) | |
Monoclonal | |
0.2mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
25 μg | |
Cell Biology | |
Mouse | |
Purified |
Flow Cytometry | |
10B4 (also known as 24/04-10B4) | |
Brilliant Blue 515 (BB515) | |
CD370; CLC9A; Clec9a; DNGR-1; C-type lectin domain family 9 member A | |
Mouse Clec9A Peptide | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed. | |
IgG2a κ |
For Research Use Only.