Learn More
CD31/PECAM-1 Rat anti-Porcine, Clone: 377537, R&D Systems™
GREENER_CHOICE
Rat Monoclonal Antibody has been used in 4 publications
Brand:  R&D Systems MAB33871-SP
Encompass
Product Code. 15766194
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
CD31/PECAM-1 Monoclonal specifically detects CD31/PECAM-1 in Porcine samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.