Learn More
CD24 Mouse anti-Human, PE, Clone: ML5, BD
Mouse Monoclonal Antibody
Brand: BD Biosciences 555428
Additional Details : Weight : 0.02000kg
Description
The ML5 monoclonal antibody specifically binds to CD24. CD24 is a 35-45 kDa two-chain glycoprotein expressed on the surface of B cells, granulocytes and most B-cell lines. CD24 may play a role in the regulation of B-cell proliferation and maturation.
Specifications
CD24 | |
Monoclonal | |
PE | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
ML5 | |
Heat Stable Antigen Homologue (HSA); Ba-1; CD24A | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG2a κ |
For Research Use Only.