Learn More
CD2 Mouse anti-Human, PE-CF594, Clone: RPA-2.10, BD
Mouse Monoclonal Antibody
280.00€ - 660.00€
Specifications
Antigen | CD2 |
---|---|
Clone | RPA-2.10 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | PE-CF594 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15811819
|
BD Biosciences
562319 |
25 Tests |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
15821799
|
BD Biosciences
562300 |
100 Tests |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
||||||
Description
The RPA-2.10 monoclonal antibody specifically binds to CD2. CD2 is a 50 kDa single-chain transmembrane glycoprotein, also known as LFA-2 or the receptor for sheep erythrocytes. CD2 belongs to the immunoglobulin superfamily of proteins along with its ligand LFA-3, CD58. It is present on about 80-90% of human peripheral blood lymphocytes, greater than 95% of thymocytes, all T lymphocytes that form E-rosettes and a subset of NK cells. CD2 plays a role in T-cell signaling and in lymphocyte adhesion.
This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red (eg 610/20-nm filter).
Specifications
CD2 | |
Flow Cytometry | |
PE-CF594 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
IgG1 κ | |
Affinity Purified | |
LFA-2; Lymphocyte function antigen-2; T11/Leu-5; Rosette receptor |
RPA-2.10 | |
Monoclonal | |
Mouse | |
Human | |
Human Phytohemagglutinin-treated Lymphoblasts | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.