missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD185 (CXCR5) Rat anti-Mouse, BB700, Clone: 2G8, BD Horizon™
Rat Monoclonal Antibody
279.00€ - 655.00€
Specifications
Antigen | CD185 (CXCR5) |
---|---|
Clone | 2G8 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15835099
|
BD Biosciences
566493 |
25 μg |
279.00€
Each |
Estimated Shipment: 21-05-2024
Log in to see stock. |
|||||
15825099
|
BD Biosciences
566492 |
0.1 mg |
655.00€
Each |
Estimated Shipment: 23-05-2024
Log in to see stock. |
|||||
Description
CD185 (CXCR5) Monoclonal antibody specifically detects CD185 (CXCR5) in Mouse samples. It is validated for Flow Cytometry.Specifications
CD185 (CXCR5) | |
0.2 mg/mL | |
Monoclonal | |
Purified | |
RUO | |
IgG2a κ | |
Refrigerate (2 to 8°C) |
2G8 | |
Flow Cytometry | |
Brilliant Blue 700 (BB700) | |
Rat | |
Mouse | |
Primary |