missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CCT 369260
GREENER_CHOICE

Product Code. 30000443
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 30000443

Brand: Tocris Bioscience™ 7799/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Molecular glue; induces degradation of BCL6

CCT 369260 acts as a potent BCL6 molecular glue Degrader (IC50 = 54 nM). It reduces BCL6 levels in a lymphoma xenograft mouse model. CCT 369260 is orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material rel-5-[[5-Chloro-2-[(3R,5S)-4,4-difluoro-3,5-dimethyl-1-piperidinyl]-4-pyrimidinyl]amino]-1,3-dihydro-3-(3-hydroxy-3-methylbutyl)-1-methyl-2H-benzimidazol-2-one
CAS 2253878-44-1
Quantity 25 mg
Target Molecular Glues
Molecular Formula C24H31ClF2N6O2
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.