missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CCT 369260

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
CCT 369260 acts as a potent BCL6 molecular glue Degrader (IC50 = 54 nM). It reduces BCL6 levels in a lymphoma xenograft mouse model. CCT 369260 is orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | rel-5-[[5-Chloro-2-[(3R,5S)-4,4-difluoro-3,5-dimethyl-1-piperidinyl]-4-pyrimidinyl]amino]-1,3-dihydro-3-(3-hydroxy-3-methylbutyl)-1-methyl-2H-benzimidazol-2-one |
| CAS | 2253878-44-1 |
| Quantity | 5 mg |
| Target | Molecular Glues |
| Molecular Formula | C24H31ClF2N6O2 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction