Learn More
Motic™ CCIS™ Plan Achromatic Strain-free Objectives
Product Code. 17198618
Click to view available options
Magnification Power:
10X
20X
40X
4X
60X
Numerical Aperture:
0.1
0.25
0.4
0.65
0.8
Working Distance:
0.35 mm
0.6 mm
0.8 mm
17.4 mm
30.5 mm
Unit Size:
Each
Product Code. 17198618
Brand: Motic™ 1101001707191
Only null left
Add to basket
Add to basket
Plan Achromatic strain-free objectives for use with TEC POL Microscopes.
  • For use with Motic™ Panthera TEC POL Microscopes
  • UC PL P Plan Achromatic strain-free objectives
Specifications
Type Microscope Objective
For Use With (Equipment) Motic™ TEC POL Microscopes
Lens Type Plan Achromatic
Magnification Power 4X
Numerical Aperture 0.1
Working Distance 30.5 mm