Learn More
Novus Biologicals™ CCDC138 293T Cell Transient Overexpression Lysate
Product Code. 18596301 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18596301 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 18596301 Supplier Novus Biologicals™ Supplier No. H00165055T01
 Restricted Item
Estimated Shipment: 17-08-2026
Specifications
Quantity 0.1 mL
Content And Storage Store at -80°C. Avoid freeze/thaw cycles.
Target CCDC138
Synonym coiled-coil domain containing 138, coiled-coil domain-containing protein 138, FLJ32745
Gene Symbol CCDC138