Learn More
CBS Scientific Gradient Markers
Product Code. 11764696
Change view
Click to view available options
Volume (Metric):
50 mL
250 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
11764696 250 mL Each
11789545 50 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11764696 Supplier CBS Scientific Supplier No. GM500
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Linear with side outlets.
  • Gradient flow is controlled by a PTFE-coated valve centered between the two chambers
  • The outflow channel is fitted with a male luer adapter which can be used with any needle or luer valve
Specifications
For Use With (Application) Electrophoresis
Type Gradient Marker
Volume (English) 8.45 oz.
Volume (Metric) 250 mL