missing translation for 'onlineSavingsMsg'
Learn More

CBP,EIF4E Rabbit anti-Human, CoraLite 488, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

129.00€ - 430.00€

Specifications

Antigen CBP,EIF4E
Concentration 1000 μg/mL
Applications Flow Cytometry
Classification Polyclonal
Conjugate CoraLite 488
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17731249
View Documents Promotion Details
Proteintech
CL488-11149-20UL
20 μL
129.00€
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
 
17721249
View Documents Promotion Details
Proteintech
CL488-11149-100UL
100 μL
430.00€
100µL
View alternative products
Save Up to 
»
Estimated Shipment: 18-06-2024
to see stock.
Add to basket
 
Description

Description

CBP,EIF4E Polyclonal antibody specifically detects CBP,EIF4E in Human samples. It is validated for FC (Intra)
Specifications

Specifications

CBP,EIF4E
Flow Cytometry
CoraLite 488
Rabbit
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
1977
Primary
Store at -20°C
1000 μg/mL
Polyclonal
Liquid
Human
BC020968
IgG
Antigen affinity purification
Antibody
Videos
SDS
Documents

Documents

Product Certifications