missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CBMR 0083
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CBMR 0083 is a potent allosteric free fatty acid receptor 1 (FFA1) and 4 (FFA4) agonist (EC50 values are 1 and 2 nM for FFA1 and FFA4, respectively).
Specifications
Specifications
| Chemical Name or Material | 1-[3-[[4-Amino-6-[[2-(2,5-dichlorophenyl)ethyl]amino]-1,3,5-triazin-2-yl]methoxy]-5-methylphenyl]-2-pyrrolidinone |
| CAS | 2839486-16-5 |
| Quantity | 50 mg |
| Target | Free Fatty Acid Receptor Agonists |
| Molecular Formula | C23H24Cl2N6O2 |
| Purity | 0.98 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction