Learn More
Fisherbrand™ Catalyst Platinum-Cured Silicone Formulation Pump Tubing
Product Code. p-6877017 Shop All Fisherbrand Products
Click to view available options
Diameter (Metric) Inner:
4.8 mm
6.4 mm
8 mm
9.5 mm
Thickness (Metric):
1.6 mm
2.4 mm
Unit Size:
Each
Product Code. 15561315
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Available in standard 15m Lengths.
    • Includes platinum-cured silicone, two TPEs and long-life PVC tubing material
    • Available in both thin and thick wall configurations
Specifications
Description Catalyst pump tubing
Diameter (English) Inner 0.19 in.
Diameter (Metric) Inner 4.8 mm
Material Solva PVC
Overall Length (English) 49.2 ft.
Overall Length (Metric) 15 m
Thickness (English) 0.06 in.
Thickness (Metric) 1.6 mm
Product Type Pump Tubing